Methylblau, Sigma-Aldrich®

Lieferant: SIGMA ALDRICH MICROSCOPY

Synonyme: Baumwollblau, Acid Blue 93, Anilinblau WS, Wasserblau 6B, Aniline blue water soluble, Anilinblau wasserlöslich, Aniline blue WS, Dinatrium-[[4-[bis[4-[(sulfonatophenyl)amino]phenyl]methylen]cyclohexa-2,5-dien-1-yliden]amino]benzolsulfonat

M-Clarity™ Qualitätsstufe = MQ200

95290-100G 95290-25G 95290-5G
95290-100GEA 289 EUR
95290-100G 95290-25G 95290-5G
Methylblau, Sigma-Aldrich®
Methylblau

Methyl blue (C.l. 42780) is a triaminotriphenylmethane dye with each amino group carrying a monosulfonated phenyl substituent. It is also known as Water blue, Acid blue 93, Cotton blue, Poirriers blue, and Aniline blue water soluble.


  • Methyl blue has been used to stain leaf disks for callose visualisation within the cells
  • It has been used to incubate cells for 4',6-diamidino-2-phenylindole (DAPI)/septa staining
  • Used as a marker for β-(1-3)-d-glucans or acidic phloroglucinol for the detection of callose and lignin
  • Used in Mann's stain as it involves a methyl blue-eosin mixture

Formel: C₃₇H₂₇N₃Na₂O₉S₃
Flammpunkt: >300 °C (Pensky-Martens) (geschlossener Tiegel)
Lagertemperatur: Raumtemperatur
MDL: MFCD00058509
CAS-Nummer: 28983-56-4
EINECS: 249-352-9
Merck Index: 13,06055
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99c81fa4222b5d619a577fa4fe452d01b0484cb6&vid=e8725a21d295d576c06aa9d0f7684176aa5dd095&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Appearance (colour) Purple to very dark purple
Appearance (form) Powder or crystals
Thin layer chromatogr. Corresponds to standard
Solubility (colour) 0.01 g in 10 ml water
Solubility (method) 0.01 g in 10 ml water
pH-transition 14.0
Colour Pink
Wavelength (1) (UV) 595.0 - 605.0 nm
Extinction (1%/1 cm) (1) (UV) ≥400
Wavelength (2) (UV) 310.0 - 320.0 nm
Extinction (1%/1 cm) (2) (UV) ≥200
Solvent (UV) HCl 0.005 m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR