Methylblau, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
Baumwollblau, Acid Blue 93, Anilinblau WS, Wasserblau 6B, Aniline blue water soluble, Anilinblau wasserlöslich, Aniline blue WS, Dinatrium-[[4-[bis[4-[(sulfonatophenyl)amino]phenyl]methylen]cyclohexa-2,5-dien-1-yliden]amino]benzolsulfonat
M-Clarity™ Qualitätsstufe = MQ200
95290-100G
95290-25G
95290-5G
95290-100GEA
289
EUR
95290-100G
95290-25G
95290-5G
Methylblau, Sigma-Aldrich®
Methylblau
Methyl blue (C.l. 42780) is a triaminotriphenylmethane dye with each amino group carrying a monosulfonated phenyl substituent. It is also known as Water blue, Acid blue 93, Cotton blue, Poirriers blue, and Aniline blue water soluble.
- Methyl blue has been used to stain leaf disks for callose visualisation within the cells
- It has been used to incubate cells for 4',6-diamidino-2-phenylindole (DAPI)/septa staining
- Used as a marker for β-(1-3)-d-glucans or acidic phloroglucinol for the detection of callose and lignin
- Used in Mann's stain as it involves a methyl blue-eosin mixture
Formel:
C₃₇H₂₇N₃Na₂O₉S₃ Flammpunkt: >300 °C (Pensky-Martens) (geschlossener Tiegel) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00058509 CAS-Nummer: 28983-56-4 EINECS: 249-352-9 Merck Index: 13,06055 |
|
Spezifikation Testergebnisse
Appearance (colour) | Purple to very dark purple |
Appearance (form) | Powder or crystals |
Thin layer chromatogr. | Corresponds to standard |
Solubility (colour) | 0.01 g in 10 ml water |
Solubility (method) | 0.01 g in 10 ml water |
pH-transition | 14.0 |
Colour | Pink |
Wavelength (1) (UV) | 595.0 - 605.0 nm |
Extinction (1%/1 cm) (1) (UV) | ≥400 |
Wavelength (2) (UV) | 310.0 - 320.0 nm |
Extinction (1%/1 cm) (2) (UV) | ≥200 |
Solvent (UV) | HCl 0.005 m |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...