Wasser, TraceSELECT™ Trace_analysis, Riedel-de Haën™
Lieferant: Honeywell Chemicals
Synonyme:
Wasserstoffoxid, Hydrogenoxid
95305-1L
95305-2.5L
95305-500ML
95305-4X2.5L
95305-10L
95305-250ML
95305-1LEA
59.81
EUR
95305-1L
95305-2.5L
95305-500ML
95305-4X2.5L
95305-10L
95305-250ML
Wasser, TraceSELECT™ Trace_analysis, Riedel-de Haën™
Wasser
High purity solvents for speciation analysis by LC-ICP-MS. Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance. The blank values for metal traces in these solvents are in the ppb range or lower.
Formel:
H₂O Molecular Weight: 18.02 g/mol Siedepunkt: 100 °C (1.013 hPa) Schmelzpunkt: 0 °C Dichte: 1 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011332 CAS-Nummer: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Spezifikation Testergebnisse
Conductivity 25 °C | Max. 2.0 µS/cm |
Non-volatile matter | Max. 1 ppm |
Silver (Ag) | Max. 1 ppb |
Aluminium (Al) | Max. 1 ppb |
Arsenic (As) | Max. 1 ppb |
Gold (Au) | Max. 1 ppb |
Barium (Ba) | Max. 1 ppb |
Beryllium (Be) | Max. 1 ppb |
Bismuth (Bi) | Max. 1 ppb |
Calcium (Ca) | Max. 5 ppb |
Cadmium (Cd) | Max. 1 ppb |
Cerium (Ce) | Max. 1 ppb |
Cobalt (Co) | Max. 1 ppb |
Chromium (Cr) | Max. 1 ppb |
Cesium (Cs) | Max. 1 ppb |
Copper (Cu) | Max. 1 ppb |
Iron (Fe) | Max. 5 ppb |
Gallium (Ga) | Max. 1 ppb |
Germanium (Ge) | Max. 1 ppb |
Mercury (Hg) | Max. 1 ppb |
Indium (In) | Max. 1 ppb |
Iridium (Ir) | Max. 1 ppb |
Potassium (K) | Max. 5 ppb |
Lithium (Li) | Max. 1 ppb |
Magnesium (Mg) | Max. 1 ppb |
Manganese (Mn) | Max. 1 ppb |
Molybdenum (Mo) | Max. 1 ppb |
Sodium (Na) | Max. 5 ppb |
Nickel (Ni) | Max. 1 ppb |
Lead (Pb) | Max. 1 ppb |
Palladium (Pd) | Max. 1 ppb |
Platinum (Pt) | Max. 1 ppb |
Rubidium (Rb) | Max. 1 ppb |
Rhodium (Rh) | Max. 1 ppb |
Ruthenium (Ru) | Max. 1 ppb |
Antimony (Sb) | Max. 1 ppb |
Selenium (Se) | Max. 1 ppb |
Tin (Sn) | Max. 1 ppb |
Strontium (Sr) | Max. 1 ppb |
Titanium (Ti) | Max. 1 ppb |
Thallium (Tl) | Max. 1 ppb |
Uran (U) | Max. 0.5 ppb |
Vanadium (V) | Max. 1 ppb |
Zinc (Zn) | Max. 1 ppb |
Zirconium (Zr) | Max. 1 ppb |
Silicon (as SiO₂) | Max. 0.01 ppm |
Bromide (Br) | Max. 10.0 ppb |
Chloride (Cl) | Max. 10.0 ppb |
Fluoride (F) | Max. 10.0 ppb |
Iodide (I) | Max. 10.0 ppb |
Nitrite (NO₂) | Max. 10.0 ppb |
Nitrate (NO₃) | Max. 10.0 ppb |
Phosphate (PO₄) | Max. 10.0 ppb |
Sulfate (SO₄) | Max. 10.0 ppb |
Ammonium (NH₄) | Max. 0.010 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...