Wasser, TraceSELECT™ Trace_analysis, Riedel-de Haën™

Lieferant: Honeywell Chemicals

Synonyme: Wasserstoffoxid, Hydrogenoxid

95305-1L 95305-2.5L 95305-500ML 95305-4X2.5L 95305-10L 95305-250ML
95305-1LEA 59.81 EUR
95305-1L 95305-2.5L 95305-500ML 95305-4X2.5L 95305-10L 95305-250ML
Wasser, TraceSELECT™ Trace_analysis, Riedel-de Haën™
Wasser

High purity solvents for speciation analysis by LC-ICP-MS. Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance. The blank values for metal traces in these solvents are in the ppb range or lower.

Formel: H₂O
Molecular Weight: 18.02 g/mol
Siedepunkt: 100 °C (1.013 hPa)
Schmelzpunkt: 0 °C
Dichte: 1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Conductivity 25 °C Max. 2.0 µS/cm
Non-volatile matter Max. 1 ppm
Silver (Ag) Max. 1 ppb
Aluminium (Al) Max. 1 ppb
Arsenic (As) Max. 1 ppb
Gold (Au) Max. 1 ppb
Barium (Ba) Max. 1 ppb
Beryllium (Be) Max. 1 ppb
Bismuth (Bi) Max. 1 ppb
Calcium (Ca) Max. 5 ppb
Cadmium (Cd) Max. 1 ppb
Cerium (Ce) Max. 1 ppb
Cobalt (Co) Max. 1 ppb
Chromium (Cr) Max. 1 ppb
Cesium (Cs) Max. 1 ppb
Copper (Cu) Max. 1 ppb
Iron (Fe) Max. 5 ppb
Gallium (Ga) Max. 1 ppb
Germanium (Ge) Max. 1 ppb
Mercury (Hg) Max. 1 ppb
Indium (In) Max. 1 ppb
Iridium (Ir) Max. 1 ppb
Potassium (K) Max. 5 ppb
Lithium (Li) Max. 1 ppb
Magnesium (Mg) Max. 1 ppb
Manganese (Mn) Max. 1 ppb
Molybdenum (Mo) Max. 1 ppb
Sodium (Na) Max. 5 ppb
Nickel (Ni) Max. 1 ppb
Lead (Pb) Max. 1 ppb
Palladium (Pd) Max. 1 ppb
Platinum (Pt) Max. 1 ppb
Rubidium (Rb) Max. 1 ppb
Rhodium (Rh) Max. 1 ppb
Ruthenium (Ru) Max. 1 ppb
Antimony (Sb) Max. 1 ppb
Selenium (Se) Max. 1 ppb
Tin (Sn) Max. 1 ppb
Strontium (Sr) Max. 1 ppb
Titanium (Ti) Max. 1 ppb
Thallium (Tl) Max. 1 ppb
Uran (U) Max. 0.5 ppb
Vanadium (V) Max. 1 ppb
Zinc (Zn) Max. 1 ppb
Zirconium (Zr) Max. 1 ppb
Silicon (as SiO₂) Max. 0.01 ppm
Bromide (Br) Max. 10.0 ppb
Chloride (Cl) Max. 10.0 ppb
Fluoride (F) Max. 10.0 ppb
Iodide (I) Max. 10.0 ppb
Nitrite (NO₂) Max. 10.0 ppb
Nitrate (NO₃) Max. 10.0 ppb
Phosphate (PO₄) Max. 10.0 ppb
Sulfate (SO₄) Max. 10.0 ppb
Ammonium (NH₄) Max. 0.010 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR