Organic reference standard, (-)-alpha-Bisabolol, analytical standard, Supelco®
Lieferant: Merck
95426-1ML
95426-1MLEA 88.3 EUR
95426-1ML
Organic reference standard, (-)-alpha-Bisabolol, analytical standard, Supelco®
Standards Organische Standards
(−)-α-Bisabolol is a monocyclic sesquiterpene alcohol naturally found in the essential oil of different plants like Vanillosmopsis erythropappa and Matricaria chamomilla.
It finds applications in clinical and cosmetic industries due to its pharmacological properties like anti-inflammatory, anti-irritant, antimicrobial and skin healing property. It also shows effective action against malignant tumor cells.
Verpackung: Bottomless glass bottle. Contents are inside inserted fused cone.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=39ab9d2b4614cd67b86c6f50a166cd40acfd5eaf&vid=39ab9d2b4614cd67b86c6f50a166cd40acfd5eaf&prid=web&clid=SAPDAM