Micro particles based on melamine resin nile blue-marked, analytical standard, Supelco®
Lieferant: Merck
95611-10ML-F
95611-10ML-FEA
707
EUR
95611-10ML-F
Micro particles based on melamine resin nile blue-marked, analytical standard, Supelco®
Standards
Partikelgrößenanalyse-Standards
For every lot, exact values of particle size and standard deviation are determined with an accuracy of 0,01 μm using a Coulter Multisizer.
Fluorescence: λex 633 nm; λem 672 nm