Apiezon® Wachs W 40
Lieferant: M I MATERIALS
FAPW400250
96030.340EA 371 EUR
96030.340
Apiezon® Wachs W 40
Apiezon® Wachs W
This is the softest wax (designed to flow at temperatures of 40-50°C) and suitable for situations where the sealing medium is required to flow into or around a joint.