Citrate standard for IC, TraceCERT®, Supelco®
Lieferant: Merck
96068-100ML
96068-100MLEA 58.2 EUR
96068-100ML
Citrate standard for IC, TraceCERT®, Supelco®
Standards Anionische Standards Citrat-Standards
Standards: This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.