Salpetersäure 69% NF, J.T.Baker®

Lieferant: Avantor
Gefahr

9607-04EA 101.46 EUR
9607-04 9607-33
Salpetersäure 69% NF, J.T.Baker®
Salpetersäure
Formel: HNO₃
Siedepunkt: 120,5 °C (1013 hPa)
Schmelzpunkt: –42 °C
Dichte: 1,4 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011349
CAS-Nummer: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8,II
Merck Index: 13,06608

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 69.0 - 70.0 %
NF - Assay (as HNO₃) 69.0 - 70.0 %
NF - Clarity and Color Passes Test
NF - Identification Passes Test
NF - Chloride (Cl) ≤0.5 ppm
NF - Sulfate (SO₄) ≤1 ppm
NF - Iron (Fe) ≤0.2 ppm
NF - Residue on Ignition ≤5 ppm
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232) - Information on elemental impurities for
this product is available on the associated Product Regulatory Data
Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR