Triethylammoniumchlorid ≥99,0% für die HPLC, Fluka™
Lieferant: Honeywell Chemicals
|
Achtung
|
Synonyme:
Triethylamin Hydrochlorid
96249-250G
96249-250GEA
524.57
EUR
96249-250G
Triethylammoniumchlorid ≥99,0% für die HPLC, Fluka™
Triethylammonium chloride
Formel:
C₆H₁₆ClN Molecular Weight: 137,65 g/mol Dichte: 0,7637 g/cm³ (20 °C) Lagertemperatur: Room Temperature |
MDL:
MFCD00012500 CAS-Nummer: 554-68-7 EINECS: 209-067-2 |
Spezifikation Testergebnisse
Identity (NMR) | complying |
Assay | min. 99.0 % |
Colour of the solution | complying |
Clearness of the solution | complying |
Absorbance 500 nm | max. 0.02 |
Absorbance at 260 nm | max. 0.02 |
Absorbance at 250 nm | max. 0.03 |
Absorbance at 230 nm | max. 0.04 |
Absorbance at 220 nm | max. 0.05 |
Suitability test (HPLC) | complying |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...