Transition metal mix 2 for ICP, 8 elements, TraceCERT®, Supelco®
Lieferant: Merck
96442-100ML
96442-100MLEA
186
EUR
96442-100ML
Transition metal mix 2 for ICP, 8 elements, TraceCERT®, Supelco®
Standards
Kationischer Standards
Mehrelement-Standards
100 mg/l each: Ti, Zr, Hf, Nb, Ta, Mo, W and Re in 2% nitric acid and 1% hydrofluoric acid
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034