Zinkoxid ≥99,0% (durch komplexometrische Analyse), Puriss. p.a. ACS, Fluka™
Lieferant: Honeywell Chemicals
Synonyme:
Zink(II)oxid
96479-500G
96479-500GEA
67.08
EUR
96479-500G
Zinkoxid ≥99,0% (durch komplexometrische Analyse), Puriss. p.a. ACS, Fluka™
Zinkoxid
Formel:
ZnO Schmelzpunkt: 1,975 °C Dichte: 5,6 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011300 CAS-Nummer: 1314-13-2 EINECS: 215-222-5 REACH: 01-2119463881-32 Merck Index: 13,10200 |
Spezifikation Testergebnisse
Assay | 99.0 - 101.0 % |
Insoluble in acid | Max. 0.01 % |
Alkalinely reacting substances | complying |
Arsenic (As) | Max. 1 ppm |
Calcium (Ca) | Max. 10 ppm |
Cadmium (Cd) | Max. 5 ppm |
Cobalt (Co) | Max. 5 ppm |
Chromium (Cr) | Max. 5 ppm |
Copper (Cu) | Max. 5 ppm |
Iron (Fe) | Max. 5 ppm |
Potassium (K) | Max. 50 ppm |
Magnesium (Mg) | Max. 5 ppm |
Manganese (Mn) | Max. 5 ppm |
Sodium (Na) | Max. 10 ppm |
Nickel (Ni) | Max. 5 ppm |
Lead (Pb) | Max. 20 ppm |
Chloride (Cl) | Max. 10 ppm |
Nitrate (NO₃) | Max. 30 ppm |
Total S (as SO₄) | Max. 100 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...