Supelco® TraceCERT® Organic Reference Standard, Isobutyric acid
Lieferant: Merck
96499-100MG
96499-100MGEA
120
EUR
96499-100MG
Supelco® TraceCERT® Organic Reference Standard, Isobutyric acid
Standards
Organische Standards
- GHS signal: GHS02, GHS06, GHS05
Standards: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Bitte beachten P210 - P233 - P280 - P301 + P312 - P303 + P361 + P353 - P305 + P351 + P338