Schwefelsäure 95,0-98,0% NF, Multikompendial, J.T.Baker®

Lieferant: Avantor
Gefahr


Veränderungsmanagement-Kategorie= R

9671-02EA 100.06 EUR
9671-02 9671-03 9671-26
Schwefelsäure 95,0-98,0% NF, Multikompendial, J.T.Baker®
Schwefelsäure

GMP Manufactured Product Meets B.P. Chemical Specifications Meets E.P. Chemical Specifications Meets N.F. Requirements

Formel: H₂SO₄
Molecular Weight: 98.08 g/mol
Siedepunkt: ∼335 °C (1013 hPa)
Schmelzpunkt: 3 °C
Dichte: 1.84 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064589
CAS-Nummer: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II
Merck Index: 14,08974

Bestellen

Spezifikation Testergebnisse

GMP Manufactured Product
Meets N.F. Requirements
Meets E.P. Chemical Specifications
Meets B.P. Chemical Specifications
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical Passes Test
NF - Identification Passes Test
NF - Residue on Ignition ≤ 0.005 %
NF - Chloride (Cl) ≤ 0.005 %
NF - Arsenic (As) ≤ 1 ppm
NF - Reducing Substances Passes Test
NF - Assay (as H₂SO₄) (by acidimetry) 95.0 - 98.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Chloride (Cl) ≤ 50 ppm
EP/BP - Nitrate (NO₃) Passes Test
EP/BP - Iron (Fe) ≤ 25 ppm
EP/BP - Assay (H₂SO₄) 95.0 - 100.5 %
No Class 1,2,3 or other solvents are used or produced in the = 5 ppm
manufacturing or purification of the product. 95.0 - 100.5 %
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR