Schwefelsäure 96%, CMOS für Mikroelektronik, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
9674-06EA
1140
EUR
9674-06
9674-09
9674-26
Schwefelsäure 96%, CMOS für Mikroelektronik, J.T.Baker®
Schwefelsäure
Formel:
H₂SO₄ Molecular Weight: 98.08 g/mol Siedepunkt: ∼335 °C (1013 hPa) Schmelzpunkt: 3 °C Dichte: 1.84 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064589 CAS-Nummer: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8 II Merck Index: 14,08974 |
Spezifikation Testergebnisse
For Microelectronic Use | 95.0 - 97.0 % |
Recommended Storage Conditions: 15° - 100°F |
|
Assay (H₂SO₄) | 95.0 - 97.0 % |
Color (APHA) | ≤10 |
Residue after Ignition | ≤2 ppm |
Chloride (Cl) | ≤0.1 ppm |
Nitrate (NO₃) | ≤0.2 ppm |
Phosphate (PO₄) | ≤0.3 ppm |
Trace Impurities - Aluminum (Al) | ≤50.0 ppb |
Arsenic and Antimony (as As) | ≤5.0 ppb |
Trace Impurities - Barium (Ba) | ≤10.0 ppb |
Trace Impurities - Beryllium (Be) | ≤10.0 ppb |
Trace Impurities - Bismuth (Bi) | ≤20.0 ppb |
Trace Impurities - Boron (B) | ≤10.0 ppb |
Trace Impurities - Cadmium (Cd) | ≤10.0 ppb |
Trace Impurities - Calcium (Ca) | ≤50.0 ppb |
Trace Impurities - Chromium (Cr) | ≤50 ppb |
Trace Impurities - Cobalt (Co) | ≤10.0 ppb |
Trace Impurities - Copper (Cu) | ≤10.0 ppb |
Trace Impurities - Gallium (Ga) | ≤10.0 ppb |
Trace Impurities - Germanium (Ge) | ≤10.0 ppb |
Trace Impurities - Gold (Au) | ≤20 ppb |
Heavy Metals (as Pb) | ≤200.0 ppb |
Trace Impurities - Iron (Fe) | ≤100.0 ppb |
Trace Impurities - Lead (Pb) | ≤20.0 ppb |
Trace Impurities - Lithium (Li) | ≤10.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤50.0 ppb |
Trace Impurities - Manganese (Mn) | ≤10.0 ppb |
Trace Impurities - Mercury (Hg) | ≤5.0 ppb |
Trace Impurities - Molybdenum (Mo) | ≤10.0 ppb |
Trace Impurities - Nickel (Ni) | ≤10.0 ppb |
Trace Impurities - Niobium (Nb) | ≤10.0 ppb |
Trace Impurities - Potassium (K) | ≤50 ppb |
Trace Impurities - Silicon (Si) | ≤50 ppb |
Trace Impurities - Silver (Ag) | ≤10.0 ppb |
Trace Impurities - Sodium (Na) | ≤100.0 ppb |
Trace Impurities - Strontium (Sr) | ≤10.0 ppb |
Trace Impurities - Tantalum (Ta) | ≤10.0 ppb |
Trace Impurities - Thallium (Tl) | ≤20.0 ppb |
Trace Impurities - Tin (Sn) | ≤50 ppb |
Trace Impurities - Titanium (Ti) | ≤10.0 ppb |
Trace Impurities - Vanadium (V) | ≤10.0 ppb |
Trace Impurities - Zinc (Zn) | ≤50 ppb |
Trace Impurities - Zirconium (Zr) | ≤10.0 ppb |
Particle Count at point of fill - 0.5 µm and greater | ≤150 par/ml |
Particle Count at point of fill - 1.0 µm and greater | ≤25 par/ml |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...