Schwefelsäure 95,0-98,0%, BAKER ANALYZED® ACS, J.T.Baker®
Lieferant: Avantor
|
Gefahr
|
9681-02EA
86.57
EUR
9681-02
9681-07
9681-08
9681-15
Schwefelsäure 95,0-98,0%, BAKER ANALYZED® ACS, J.T.Baker®
Schwefelsäure
The quality of BAKER ANALYZED™ ACS reagents meets or exceeds the requirements set forth by the American Chemical Society (ACS). These acids are tested for common trace metals.
Formel:
H₂SO₄ Molecular Weight: 98.08 g/mol Siedepunkt: ∼335 °C (1013 hPa) Schmelzpunkt: 10,38 Dichte: 1.84 g/cm³ (20 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064589 CAS-Nummer: 7664-93-9 EINECS: 231-639-5 UN: 1830 ADR: 8 II |
Spezifikation Testergebnisse
Meets ACS Reagent Chemical Requirements | |
For Laboratory,Research,or Manufacturing Use | 95.0 - 98.0 % |
Assay (H₂SO₄) | 95.0 - 98.0 % |
Appearance | Passes Test |
Color (APHA) | ≤10 |
Specific Gravity at 60°/60°F | ≥1.84 |
Residue after Ignition | ≤4 ppm |
Ammonium (NH₄) | ≤1 ppm |
Trace Impurities - Arsenic (As) | ≤0.004 ppm |
Chloride (Cl) | ≤0.1 ppm |
Nitrate (NO₃) | ≤0.5 ppm |
Trace Impurities - Aluminum (Al) | ≤200.0 ppb |
Trace Impurities - Calcium (Ca) | ≤200.0 ppb |
Trace Impurities - Chromium (Cr) | ≤100.0 ppb |
Trace Impurities - Copper (Cu) | ≤100.0 ppb |
Trace Impurities - Gold (Au) | ≤100.0 ppb |
Heavy Metals (as Pb) | ≤500.0 ppb |
Trace Impurities - Iron (Fe) | ≤200.0 ppb |
Trace Impurities - Lead (Pb) | ≤200.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤200.0 ppb |
Trace Impurities - Manganese (Mn) | ≤100.0 ppb |
Trace Impurities - Mercury (Hg) | ≤5.0 ppb |
Trace Impurities - Nickel (Ni) | ≤200.0 ppb |
Trace Impurities - Potassium (K) | ≤300.0 ppb |
Trace Impurities - Sodium (Na) | ≤300 ppb |
Trace Impurities - Tin (Sn) | ≤200.0 ppb |
Trace Impurities - Titanium (Ti) | ≤200.0 ppb |
Trace Impurities - Zinc (Zn) | ≤200.0 ppb |
Meets Reagent Specifications for testing USP/NF monographs |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...