Organic reference standard, Benzylidene camphor sulphonic acid, analytical standard, Supelco®
Lieferant: Merck
97085-25MG
97085-25MGEA 153 EUR
97085-25MG
Organic reference standard, Benzylidene camphor sulphonic acid, analytical standard, Supelco®
Standards Organische Standards
Benzylidene camphor sulphonic acid (BCSA) is an organic ultraviolet absorbing compound.
  • Used primarily in cosmetics (sunscreens, lipsticks, shampoos and hair sprays)
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83071df945f7c56908e76eec637db277c341dee5&vid=a76652406b14db3d9ec159917d274c4fbd4c07b7&prid=web&clid=SAPDAM