Organic reference standard, Benzylidene camphor sulphonic acid, analytical standard, Supelco®
Lieferant: Merck
97085-25MG
97085-25MGEA
153
EUR
97085-25MG
Organic reference standard, Benzylidene camphor sulphonic acid, analytical standard, Supelco®
Standards
Organische Standards
Benzylidene camphor sulphonic acid (BCSA) is an organic ultraviolet absorbing compound.
- Used primarily in cosmetics (sunscreens, lipsticks, shampoos and hair sprays)