Kieselgel C18-RP, Supelco®

Lieferant: Merck

Synonyme: Bulk Silica - C18, Octadecylsilicate, C18 Silica gel

97727-U
97727-UEA 423 EUR
97727-U
Kieselgel C18-RP, Supelco®
Silica gel C18-RP
Lagertemperatur: Raumtemperatur
MDL: MFCD00241275
CAS-Nummer: 71889-02-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR