Flash Silica, dried, spherical, Supelco®

Lieferant: Merck

Synonyme: Silica gel

97729-U
97729-UEA 7360 EUR
97729-U
Flash Silica, dried, spherical, Supelco®
Flash Silica
Formel: SiO₂
Molecular Weight: 60.08 g/mol
CAS-Nummer: 112926-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da04c5c4b58dc8f670d66bdcf321c463de9ca37a&vid=37ed2d40135060d69cb862b5a0263f9e36b04d42&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR