Trifluoressigsäure, BAKER ANALYZED® für die LC-MS, J.T.Baker®

Lieferant: Avantor
Gefahr

9810.0002AMEA 62.53 EUR
9810.0002AM 9810.0010AM
Trifluoressigsäure, BAKER ANALYZED® für die LC-MS, J.T.Baker®
Trifluoressigsäure
Formel: C₂HF₃O₂
Siedepunkt: 71,8 °C (1013 hPa)
Schmelzpunkt: –15 °C
Dichte: 1,490 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004169
CAS-Nummer: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 13,09754
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9501f3ac85d66e8d19d1cd1c3fe5409b974e9d8a&vid=be6aa627129b49ccda76006e7003b46d79b933cc&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay min. 99.5 %
Residue after Evaporation max. 10 ppm
LC-Gradient with Diode Array Detection (AU), 0.1% (v/v) aqueous solution:
at 220 nm max. 0.002
at 254 nm max. 0.01
LC-MS Gradient Suitability Test (TIC, 100 to 2000 m/z), 0.1% (v/v) aqueous solution:
Positive ESI-MS Sensitive Impurities (as Reserpine) max. 50 ppb
Trace Impurities (in ppb), 0.1% (v/v) aqueous solution
Aluminium (Al) max. 50
Calcium (Ca) max. 50
Iron (Fe) max. 50
Potassium (K) max. 50
Magnesium (Mg) max. 50
Sodium (Na) max. 50
Ultraviolet Absorbance, 0.1% (v/v) aqueous solution (1.00-path vs water):
at 230 nm max. 0.15
UV Cut-off, nm max. 210


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR