Acetonitril, BAKER ANALYZED® für die LC-MS, J.T.Baker®

Lieferant: Avantor
Gefahr

9821.1000GLEA 231 EUR
9821.1000GL 9821.2500GL
Acetonitril, BAKER ANALYZED® für die LC-MS, J.T.Baker®
Acetonitril
Formel: H₃CCN
Siedepunkt: 81,6 °C (1013 hPa)
Dichte: 0,782 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001878
CAS-Nummer: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3,II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

Assay (by GC) min. 99.9 %
Residue after Evaporation max. 1 ppm
Water (H2O) max. 0.01 %
LC-Gradient-Diode Array Detection (AU), test solution is modified with 0.1% (v/v) formic acid:
at 220 nm max. 0.01
at 254 nm max. 0.01
LC-MS Gradient Suitability Test (TIC, 100 to 2000 m/z), test solution is modified with 0.1% (v/v) formic acid:
Positive ESI-MS Sensitive Impurities (as Reserpine) max. 50 ng/ml
Trace Impurities (in ppb)
Aluminium (Al) max. 50
Calcium (Ca) max. 50
Iron (Fe) max. 50
Potassium (K) max. 50
Magnesium (Mg) max. 50
Sodium (Na) max. 50
Ultraviolet Absorbance (1.00-cm path vs water):
at 200 nm max. 0.05
Element concentrations are at time of lot release.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR