Wasser, BAKER ANALYZED®, J.T.Baker®

Lieferant: Avantor

9825.1000GLEA 47.58 EUR
9825.1000GL 9825.2500GL
Wasser, BAKER ANALYZED®, J.T.Baker®
Wasser
Formel: H₂O
Siedepunkt: 100 °C (1013 hPa)
Schmelzpunkt: 0 °C
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen

Spezifikation Testergebnisse

Residue after Evaporation max. 5 ppm
LC-Gradient-Diode Array Detection (AU), test solution is modified with 0.1% (v/v) formic acid:
at 210 nm max. 0.005
at 254 nm max. 0.001
LC-MS Gradient Suitability Test (TIC, 100 to 2000 m/z), test solution is modified with 0.1% (v/v) formic acid:
Positive ESI-MS Sensitive Impurities (as Reserpine) max. 100 ng/ml
Trace Impurities (in ppb)
Aluminium (Al) max. 500
Calcium (Ca) max. 100
Iron (Fe) max. 100
Potassium (K) max. 100
Magnesium (Mg) max. 100
Sodium (Na) max. 100
Element concentrations are at time of lot release.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR