Ethylacetat, BAKER ANALYZED® für die LC-MS, J.T.Baker®

Lieferant: Avantor
Gefahr

9828-03EA 295.55 EUR
9828-03
Ethylacetat, BAKER ANALYZED® für die LC-MS, J.T.Baker®
Ethylacetat
Formel: CH₃COOC₂H₅
Siedepunkt: 77,1 °C (1013 hPa)
Schmelzpunkt: –83 °C
Dichte: 0,902 g/cm³ (20 °C)
Flammpunkt: –4 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009171
CAS-Nummer: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Bestellen

Spezifikation Testergebnisse

For Laboratory,Research,or Manufacturing Use = 0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 330 - 400 nm ≤0.01
Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm ≤0.02
Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm ≤0.05
Ultraviolet Absorbance (1.00-cm cell vs. water) - 265 nm ≤0.050
Ultraviolet Absorbance (1.00-cm cell vs. water) - 255 nm ≤1.00
Assay (CH₃COOC₂H₅) ≥99.6 %
Residue after Evaporation ≤1.0 ppm
Water (H₂O) ≤0.05 %
LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine) ≤50 ppb
Trace Impurities - Aluminum (Al), For Information Only For Information Only
Trace Impurities - * Calcium (Ca), For Information Only For Information Only
Trace Impurities - Iron (Fe), For Information Only For Information Only
Trace Impurities - Magnesium (Mg), For Information Only For Information Only
Trace Impurities - Potassium (K), For Information Only For Information Only
Trace Impurities - *Sodium (Na), For Information Only
* May change over time due to extraction from glass container.
Filtered through a 0.2 micron filter.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR