Potassium ionophore I - cocktail A, Selectophore™ Material für Ionensensoren, Supelco®
Lieferant: Merck
|
Gefahr
|
Ready-to-use ion selective liquid for the preparation of microelectrodes in micropipettes
Composition:
Dibutyl sebacate, 68.0 wt. % (84838)
1,2-Dimethyl-3-nitrobenzene, 25.0 wt. % (40870)
Potassium ionophore I, 5.0 wt. % (60403)
Potassium tetrakis(4-chlorophenyl)borate, 2.0 wt. % (60591)
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH
UN:
2810 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...