Potassium ionophore I - cocktail B, Selectophore™ Material für Ionensensoren, Supelco®
Lieferant: Merck
|
Gefahr
|
Composition:
1,2-Dimethyl-3-nitrobenzene, 93.0 wt. % (40870)
Potassium ionophore I, 5.0 wt. % (60403)
Potassium tetrakis(4-chlorophenyl)borate, 2.0 wt. % (60591)
Selectophore is a trademark of Sigma-Aldrich Chemie GmbH
UN:
3172 ADR: 6.1,II |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...