VWR®, Density Standards, Certified Reference Materials, Referenced at 25 °C
Lieferant: VWR Chemicals
99656.600EA
85.7
EUR
99656.600
99660.600
VWR®, Density Standards, Certified Reference Materials, Referenced at 25 °C
Standards
Dichtestandards
VWR® ISO 17025 / ISO 17034 dual certified density standards are for the verification of instruments used to measure density of materials which are fluid at the desired test temperature.
- ISO 17025 / ISO 17034 certified under UKAS accreditation
- Density tested in strict accordance with ASTM D1480, a primary method for density determination
- Density standards range between 0,6654 - 1,6300 g/ml
- Ensures full compliance to ASTM and IP test method protocols
- Shelf life between 12 / 24 months
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...