3-Amino-3-deoxydigoxigenin hemisuccinamide, succinimidyl ester, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
A0223-5MG
A0223-5MGEA 536 EUR
A0223-5MG
3-Amino-3-deoxydigoxigenin hemisuccinamide, succinimidyl ester, Sigma-Aldrich®
3-Amino-3-deoxydigoxigenin hemisuccinamide, succinimidyl ester
Amine-reactive haptenylation reagent. This succinimidyl ester derivative of digoxigenin has been shown to inhibit the Na+/K+- ATPase by binding to the cardiac steroid receptor site.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58539305b54e3e3d6ef36233db9e61d199a3dc93&vid=1f64a239961e21b92bcd40b4440fcdc196e31c4b&prid=web&clid=SAPDAM