3-Amino-3-deoxydigoxigenin hemisuccinamide, succinimidyl ester, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
A0223-5MG
A0223-5MGEA
536
EUR
A0223-5MG
3-Amino-3-deoxydigoxigenin hemisuccinamide, succinimidyl ester, Sigma-Aldrich®
3-Amino-3-deoxydigoxigenin hemisuccinamide, succinimidyl ester
Amine-reactive haptenylation reagent. This succinimidyl ester derivative of digoxigenin has been shown to inhibit the Na+/K+- ATPase by binding to the cardiac steroid receptor site.