(2-Hydroxylpropyl)-β-cyclodextrin

Lieferant: PanReac AppliChem

Synonyme: Hydroxypropyl Betadex, Hydroxypropyl-β-cyclodextrin, (2-Hydroxypropyl)-β-cyclodextrin, HP-β-CD

A0367.0100
A0367.0100EA 312 EUR
A0367.0100
(2-Hydroxylpropyl)-β-cyclodextrin
(2-Hydroxylpropyl)-β-cyclodextrin
Formel: C₆₃H₁₁₂O₄₂
Lagertemperatur: Raumtemperatur
MDL: MFCD16621721
CAS-Nummer: 128446-35-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR