Acrylamid-Bisacrylamid (29:1) 40% aqueous solution

Lieferant: PanReac AppliChem
Gefahr

Synonyme: Acrylamid-Bis

A0385.0500 A0385.1000
A0385.0500EA 58.2 EUR
A0385.0500 A0385.1000
Acrylamid-Bisacrylamid (29:1) 40% aqueous solution
Acrylamid-Bis (32:1)
Lagertemperatur: Raumtemperatur
MDL: MFCD00080848
UN: 3426
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR