Ampicillin Natriumsalz

Lieferant: G Bioscienes
Gefahr

Synonyme: D(-)-α-Aminobenzylpenicillin Sodium Salt, Ampicillin Na, Ampicillin sodium salt, D-(−)-α-Aminobenzylpenicillin sodium salt Ampicillin sodium salt

A051-B
A051-BEA 72 EUR
A051-B
Ampicillin Natriumsalz
Ampicillin Natriumsalz
Formel: C₁₆H₁₈N₃NaO₄S
Molecular Weight: 371.39 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00064313
CAS-Nummer: 69-52-3
EINECS: 200-708-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR