Histopaque®, steril gefiltert, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Gefahr
M-Clarity™ Qualitätsstufe = MQ400
A0561-100X15ML
A0561-100X15MLEA 2470 EUR
A0561-100X15ML
Histopaque®, steril gefiltert, Sigma-Aldrich®
Histopaque®
ACCUSPIN System-Histopaque-1077 products are intended for use in the isolation of lymphocytes and other mononuclear cells. The separation medium, Histopaque-1077, is a sterile-filtered, endotoxin-tested solution of polysucrose and sodium diatrizoate, adjusted to a density of 1,077 g/ml. ACCUSPIN System-HISTOPAQUE-1077 employs γ-irradiated 50 mL polypropylene centrifuge tubes specially designed with two chambers separated by a porous high-density polyethylene barrier ('frit'). Each tube is aseptically filled with 15 ml of Histopaque 1077. Each tube will separate 15 to 30 ml of anticoagulated blood.
Bestellen
Spezifikation Testergebnisse
Appearance (Turbidity) Clear
Appearance (Form) Solution
Appearance (Colour) Colorless
Performance Conforms