n-Dodecyl-β-D-maltosid
Lieferant: PanReac AppliChem
|
Achtung
|
Synonyme:
DDM
A0819.0005
A0819.0005EA
379
EUR
A0819.0005
n-Dodecyl-β-D-maltosid
n-Dodecyl-β-D-maltosid
Formel:
C₂₄H₄₆O₁₁ Molecular Weight: 510,6 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00043012 CAS-Nummer: 69227-93-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...