Guanidiniumchlorid 8 mol/l aqueous solution
Lieferant: PanReac AppliChem
|
Achtung
|
Synonyme:
Carbamidin Hydrochlorid, Guanidin-Chlorid, Guanidinhydrochlorid, Aminomethanamidin Hydrochlorid, Aminomethansäureamidiniumchlorid
A0860.0500
A0860.0500EA
117
EUR
A0860.0500
Guanidiniumchlorid 8 mol/l aqueous solution
Guanidiniumchlorid
Formel:
NH₂C(=NH)NH₂·HCl Molecular Weight: 95.53 g/mol Schmelzpunkt: 185 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013026 CAS-Nummer: 50-01-1 EINECS: 200-002-3 Merck Index: 13,04578 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...