Cycloheximid

Lieferant: PanReac AppliChem
Gefahr

Synonyme: Naramycin A, CHX, 4-[(2R)-2-[(1S,3S,5S)-3,5-Dimethyl-2-oxocyclohexyl]-2-hydroxyethyl]piperidin-2,6-dion, Actidion

A0879.0001 A0879.0005 A0879.0025
A0879.0001EA 55.9 EUR
A0879.0001 A0879.0005 A0879.0025
Cycloheximid
Cycloheximid
Formel: C₁₅H₂₃NO₄
Molecular Weight: 281.35 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082346
CAS-Nummer: 66-81-9
EINECS: 200-636-0
UN: 2811
ADR: 6.1 I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3abdf7ad3f7b3ea9e67fba3b6f3970c6627794be&vid=6f07d0130a35bc66b22ecee29fc6c31b20823099&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR