Aluminon, Pulver, Sigma-Aldrich®
Lieferant: SIGMA ALDRICH MICROSCOPY
Synonyme:
ALuminon, Ammonium auritricarboxylate ATA Triammonium aurinetricarboxylate triammonium salt Ammonium aurintricarboxylate aurintricarboxylic acid ammonium salt Aluminon(C.1.No.43810) aurintricarboxylic acid triammonium salt, Ammonium 5,5'-((3-carboxylato-4-oxocyclohexa-2,5-dien-1-ylidene)methylene)bis(2-hydroxybenzoate), Aurin Tricarboxylic Acid Ammonium Salt
M-Clarity™ Qualitätsstufe = MQ200
Aurintricarboxylic acid ammonium salt has been used as a nuclease inhibitor to study its effects on transfection.
Aurintricarboxylic acid readily polymerises in aqueous solution, forming a stable free radical that inhibits protein-nucleic acid interactions. It is a potent inhibitor of ribonuclease and topoisomerase II by preventing the binding of the nucleic acid to the enzyme. It stimulates tyrosine phosphorylation processes including the Jak2/STAT5 pathway in NB2 lymphoma cells, ErbB4 in neuroblastoma cells, and MAP kinases, Shc proteins, phosphatidylinositide 3-kinase and phospholipase C in PC12 cells. Aurintricarboxylic acid inhibits apoptosis in many cell types. Its neuroprotective effect, perhaps due to its ability to prevent down-regulation of Ca2+ impermeable GluR2 receptors or to its ability to inhibit calpain, a Ca2+-activated protease that is activated during apoptosis.
Formel:
C₂₂H₂₃N₃O₉ Molecular Weight: 473,44 g/mol Schmelzpunkt: 220,..225 °C (Zers., Lit.) |
MDL:
MFCD00040925 CAS-Nummer: 569-58-4 EINECS: 209-319-1 |
Spezifikation Testergebnisse
Appearance (Colour) | Dark Brown to Very Dark Brown and Dark Red to Very Dark Red and Very Dark Red-Brown |
Appearance (Form) | Powder |
Solubility (Colour) 100 mg/ml in water |
Dark Red |
Water | Report Result |
Carbon Content | Report Result |
Nitrogen Content | Report Result |
Application Test Sensitivity to Aluminium Passes Test |
Corresponds to Requirements |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...