Polymyxin B-Sulfat
Lieferant: PanReac AppliChem
A0890.0001
A0890.0010
A0890.0001EA
50.5
EUR
A0890.0001
A0890.0010
Polymyxin B-Sulfat
Polymyxin B sulphate
Formel:
C₅₅H₉₆N₁₆O₁₃·2H₂SO₄ |
MDL:
MFCD00131991 CAS-Nummer: 1405-20-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...