Buffer, PBS (10X, Dulbecco's) powder pack
Lieferant: PanReac AppliChem
A0965.9010
A0965.9010EA 126 EUR
A0965.9010
Buffer, PBS (10X, Dulbecco's) powder pack
Puffer Elektrophorese-Puffer Laufpuffer
Powder mixture to prepare PBS buffer (10X) according to Dulbecco.
Composed of di-sodium hydrogen phosphate anhydrous (11500 mg/L), potassium chloride (2000 mg/L), potassium dihydrogen phosphate (2000 mg/L) and sodium chloride (80000 mg/L).