4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI)

Lieferant: PanReac AppliChem

A1001.0010 A1001.0100
A1001.0010EA 55.3 EUR
A1001.0010 A1001.0100
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI)
4',6-Diamidin-2-phenylindol-dihydrochlorid (DAPI)

DAPI (4',6-diamidine-2-phenylindole dihydrochloride) is an excellent dye for staining DNA.


DAPI is used as a reagent to detect mycoplasma or virus DNA in cell culture.

Formel: C₁₆H₁₅N₅·2HCl
Lagertemperatur: Kühlschrank
MDL: MFCD00012681
CAS-Nummer: 28718-90-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR