IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥99%, BioChemica®

Lieferant: PanReac AppliChem

A1008.0005 A1008.0025 A1008.0050 A1008.0100
A1008.0005EA 119 EUR
A1008.0005 A1008.0025 A1008.0050 A1008.0100
IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥99%, BioChemica®
IPTG (Isopropyl-β-D-thiogalactopyranosid)

Assay (HPLC): min. 99 %
α20°C/D; 1 %, H2O: -28.5° - -34.5°
pH (5 %; H2O): 5.0 - 7.0
Dioxane: max. 0.0001 %
Water (K.F.): max. 1.0 %

Bestellen

Spezifikation Testergebnisse

Assay (HPLC) min. 99 %
α20°C/D; 1 %, H2O -28.5° - -34.5°
pH (5 %; H2O) 5.0 - 7.0
Dioxane max. 0.0001 %
Water (K.F.) max. 1.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR