4-Amino-2-chlorbenzoesäure 98%

Lieferant: Thermo Scientific
Achtung

A10116.06
A10116.06EA 62.6 EUR
A10116.06
4-Amino-2-chlorbenzoesäure 98%
4-Amino-2-chlorobenzoic acid

Purity: 98% CAS No.: 2457-76-3 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: ca 220° dec. Beilstein Reference: 2803668

Formel: C₇H₆ClNO₂
Molecular Weight: 171,58 g/mol
Siedepunkt: 330 °C (1013 hPa)
Schmelzpunkt: ∼ 220 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007772
CAS-Nummer: 2457-76-3
EINECS: 219-540-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR