DL-α-Phenylalanin ≥99%

Lieferant: Thermo Fisher Scientific

Synonyme: (±)-2-Amino-3-phenylpropansäure, H-DL-Phe-OH, 3-Phenyl-DL-alanin, DL-Phenylalanin, (±)-2-Amino-3-phenylpropionic acid, (±)-2-Amino-3-phenylpropionsäure

A10132.14 A10132.22 A10132.36
A10132.14EA 28.5 EUR
A10132.14 A10132.22 A10132.36
DL-α-Phenylalanin ≥99%
DL-α-Phenylalanin

Purity: 99% CAS No.: 150-30-1 Molecular Formula: C9H11NO2 Formula Weight: 165.19 Melting Point: ca 266° dec. Beilstein Reference: 1910407 Merck Reference: 14,7271 MDL No.: MFCD00064225

Formel: C₉H₁₁NO₂
Siedepunkt: 313 °C (1013 hPa)
Schmelzpunkt: ∼ 266 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064225
CAS-Nummer: 150-30-1
EINECS: 205-756-7
Merck Index: 14,7271

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR