di-Ammoniumthiosulfat 96%

Lieferant: Thermo Fisher Scientific
Achtung

A10190.0E A10190.22 A10190.36
A10190.0EEA 108 EUR
A10190.0E A10190.22 A10190.36
di-Ammoniumthiosulfat 96%
di-Ammoniumthiosulfat

Purity: 98+% CAS No.: 7783-18-8 Molecular Formula: (NH4)2S2O3 Formula Weight: 148.20 pH: 6.0-6.5 Melting Point: ca 180° dec. MDL No.: MFCD00015965

Formel: H₈N₂O₃S₂
Schmelzpunkt: ∼ 180 °C (Zers.)
Dichte: 1,64
Lagertemperatur: Raumtemperatur
MDL: MFCD00015965
CAS-Nummer: 7783-18-8
EINECS: 231-982-0
Merck Index: 14,562

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR