L(-)-Prolin ≥99%
Lieferant: Thermo Fisher Scientific
A10199.14
A10199.22
A10199.36
A10199.14EA
36
EUR
A10199.14
A10199.22
A10199.36
L(-)-Prolin ≥99%
L(-)-Prolin
Formel:
C₅H₉NO₂ Siedepunkt: 245 °C (1013 hPa) Dichte: 1,36 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064318 CAS-Nummer: 147-85-3 EINECS: 205-702-2 Merck Index: 14,7780 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...