4-Chloranthranilsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Amino-4-chlorobenzoic acid

A10201.18 A10201.14 A10201.22 A10201.30
A10201.18EA 84.4 EUR
A10201.18 A10201.14 A10201.22 A10201.30
4-Chloranthranilsäure 98%
4-Chloranthranilsäure

Purity: 98% CAS No.: 89-77-0 Molecular Formula: C7H6ClNO2 Formula Weight: 171.58 Melting Point: ca 232° dec. Beilstein Reference: 743349

Formel: C₇H₆ClNO₂
Molecular Weight: 171.58 g/mol
Siedepunkt: 330
Schmelzpunkt: ∼ 232 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00007778
CAS-Nummer: 89-77-0
EINECS: 201-938-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR