2-Amino-3-methylpyridin 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
3-Methyl-2-pyridylamin, 3-Methylpyridin-2-amin, 2-Amino-3-picolin
A10224.14
A10224.22
A10224.36
A10224.14EA
25.2
EUR
A10224.14
A10224.22
A10224.36
2-Amino-3-methylpyridin 97%
2-Amino-3-methylpyridin
Purity: 97%
CAS No.: 1603-40-3
Molecular Formula: C6H8N2
Formula Weight: 108.14
Flash Point: 118°(244°F)
Refractive Index: 1.5825
Odour: Unpleasant
Melting Point: 29-32°
Boiling Point: 221-222°
Beilstein Reference: 107892
UN No.: 2811
MDL No.: MFCD00006320
Formel:
C₆H₈N₂ Molecular Weight: 108.14 g/mol Siedepunkt: 221…222 °C Schmelzpunkt: 29…32 °C Dichte: 1.073 Flammpunkt: 118 °C (244 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006320 CAS-Nummer: 1603-40-3 EINECS: 216-501-4 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...