2-Picolylchlorid Hydrochlorid ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A10226.14 A10226.06 A10226.22
A10226.14EA 113 EUR
A10226.14 A10226.06 A10226.22
2-Picolylchlorid Hydrochlorid ≥98%
2-Picolylchlorid Hydrochlorid

Purity: 98% CAS No.: 6959-47-3 Molecular Formula: C6H6ClN.HCl Formula Weight: 164.04 Melting Point: 120-124° Beilstein Reference: 3689155 UN No.: 3261 MDL No.: MFCD00012811

Formel: C₆H₇Cl₂N
Schmelzpunkt: 120…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012811
CAS-Nummer: 6959-47-3
EINECS: 230-149-9
UN: 3261
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR