L(-)-Tryptophan ≥99%

Lieferant: Thermo Fisher Scientific

A10230.14 A10230.22 A10230.36
A10230.14EA 47.3 EUR
A10230.14 A10230.22 A10230.36
L(-)-Tryptophan ≥99%
L(-)-Tryptophan

Purity: 99% CAS No.: 73-22-3 Molecular Formula: C11H12N2O2 Formula Weight: 204.23 Melting Point: 280° 285° Beilstein Reference: 86197 Merck Reference: 14,9797 Optical Rotation: -31° (c=1 in water) MDL No.: MFCD00064340

Formel: C₁₁H₁₂N₂O₂
Schmelzpunkt: 280…285 °C
Dichte: 1,34 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00064340
CAS-Nummer: 73-22-3
EINECS: 200-795-6
Merck Index: 14,9797

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR