Atropine sulphate monohydrate ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

A10236.09 A10236.18
A10236.09EA 50.9 EUR
A10236.09 A10236.18
Atropine sulphate monohydrate ≥98%
Atropine sulfate monohydrate

Purity: 98+% CAS No.: 5908-99-6 Molecular Formula: C34H46N2O6.H2SO4.H2O Formula Weight: 694.84 (676.82anhy) Melting Point: 187-192° Beilstein Reference: 6109275 UN No.: UN1544 MDL No.: MFCD00074815

Formel: C₁₇H₂₃NO₃
Schmelzpunkt: 187…192 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00074815
CAS-Nummer: 5908-99-6
EINECS: 200-235-0
ADR: 6.1,II
Merck Index: 14,875

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR