2,2',4,4'-Tetrahydroxybenzophenon ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Bis(2,4-dihydroxyphenyl)methanone
A10237.14
A10237.22
A10237.14EA
43.5
EUR
A10237.14
A10237.22
2,2',4,4'-Tetrahydroxybenzophenon ≥98%
2,2',4,4'-Tetrahydroxybenzophenon
Purity: 98+%
CAS No.: 131-55-5
Molecular Formula: C13H10O5
Formula Weight: 246.22
Melting Point: 198-203°
Beilstein Reference: 1914746
Formel:
C₁₃H₁₀O₅ Siedepunkt: 100…102 °C (40 mmHg) Schmelzpunkt: 198…203 °C Dichte: 1,216 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002278 CAS-Nummer: 131-55-5 EINECS: 205-028-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...