Tetrabutylammoniumbromid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

A10249.0E A10249.22 A10249.36
A10249.0EEA 703 EUR
A10249.0E A10249.22 A10249.36
Tetrabutylammoniumbromid ≥98%
Tetrabutylammoniumbromid

Purity: 98+% CAS No.: 1643-19-2 Molecular Formula: C16H36BrN Formula Weight: 322.38 Odour: Amine-like Melting Point: 101-105° Beilstein Reference: 3570983 MDL No.: MFCD00011633

Formel: C₁₆H₃₆BrN
Schmelzpunkt: 101…105 °C
Dichte: 1,15
Lagertemperatur: Raumtemperatur
MDL: MFCD00011633
CAS-Nummer: 1643-19-2
EINECS: 216-699-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR